Learn More
Waters Corp Adaptor KF25, AutoSpec Premier Mass Spectrometer
Catalog No. 50466752 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This adaptor is a KF25 component, typically used in vacuum systems to connect different parts. It is specifically designed for compatibility with certain mass spectrometer systems.
  • Compatible with AutoSpec Premier mass spectrometers.
  • Supplied as a single unit.
Catalog No. 50-466-752 Supplier Waters Corp Supplier No. S100440AD1
This adaptor is a KF25 component, typically used in vacuum systems to connect different parts. It is specifically designed for compatibility with certain mass spectrometer systems.
  • Compatible with AutoSpec Premier mass spectrometers.
  • Supplied as a single unit.