Learn More
Med Vet International Dynarex Pressure Line Adaptor, Barbed Elbow Accepts 5-7 mm Id Tubing, 22 mm Od X 22 mm Id, 50/Case
Catalog No. 502831243
Encompass_Preferred
Dynarex Resp-O2™ Connectors and Adaptors are engineered to provide improved adaptability and connectivity for use in a broad range of respiratory therapy applications—including oxygen therapy, aerosol therapy, and ventilation. These high-quality devices are manufactured from top-grade plastic materials for optimal durability and dependability. Individual models include features such as swivel, cuffed, or barbed designs for added adaptability for multiple connections. Dynarex Resp-O2™ Connectors and Adaptors are premium-grade devices designed to provide optimal connectivity and versatility for use in a wide range of respiratory therapy applications.
Catalog No. 50-283-1243 Supplier Med Vet International Supplier No. 34513
Dynarex Resp-O2™ Connectors and Adaptors are engineered to provide improved adaptability and connectivity for use in a broad range of respiratory therapy applications—including oxygen therapy, aerosol therapy, and ventilation. These high-quality devices are manufactured from top-grade plastic materials for optimal durability and dependability. Individual models include features such as swivel, cuffed, or barbed designs for added adaptability for multiple connections. Dynarex Resp-O2™ Connectors and Adaptors are premium-grade devices designed to provide optimal connectivity and versatility for use in a wide range of respiratory therapy applications.