Learn More
Glass Expansion Adaptor S19-B13-90deg
SDP
Catalog No. NC2212771
Encompass_Preferred
Optional ninety-degree transfer tube adaptor with 13mm male ball joint to spray chamber and 19mm female ball joint to Glass Expansion D-Torch. Compatible with Glass Expansion’s D-Torch for Spectro Blue EOP ICP-OES (P/N 30-808-3600).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
This item is not returnable. View return policy
Catalog No. NC2212771 Supplier Glass Expansion Supplier No. 218094048
Optional ninety-degree transfer tube adaptor with 13mm male ball joint to spray chamber and 19mm female ball joint to Glass Expansion D-Torch. Compatible with Glass Expansion’s D-Torch for Spectro Blue EOP ICP-OES (P/N 30-808-3600).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001