Learn More
Glass Expansion Adaptor S19 B19 90deg
SDP
Catalog No. 502703173
Encompass_Preferred
Adaptor S19 B19 90deg. Compatible with Glass Expansion’s D-Torch for Spectro (Ametek) ARCOS II EOP, Blue EOP and Blue TI ICP-OES. Used to as aerosol transfer tube from spray chamber to D-Torch. The following D-Torches can use this adaptor: ARCOS II EOP (30-808-3657), Blue EOP (501907034, 30-808-3600), Blue TI (501907036, 30-808-3629). Metal Ball Joint Clip is JC-19.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3173 Supplier Glass Expansion Supplier No. 218094045
Adaptor S19 B19 90deg. Compatible with Glass Expansion’s D-Torch for Spectro (Ametek) ARCOS II EOP, Blue EOP and Blue TI ICP-OES. Used to as aerosol transfer tube from spray chamber to D-Torch. The following D-Torches can use this adaptor: ARCOS II EOP (30-808-3657), Blue EOP (501907034, 30-808-3600), Blue TI (501907036, 30-808-3629). Metal Ball Joint Clip is JC-19.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001