Learn More
Starna Cells Inc ADAPTR 45M F/SQ MICROFLUORMTR
SDP
Catalog No. NC9876152
Encompass_Preferred
Adaptors for use with the Square Micro Fluorometer cells listed above, Black Anodized, Ext Width: 12.5mm, Ext Length: 12.5mm, Ext Height: 30mm, Int Width: 5.55mm, Int Length: 5.55mm, Aperature Width: 2.5mm, Aperature Height: 11mm
Catalog No. NC9876152 Supplier Starna Cells Inc Supplier No. FCA3
Adaptors for use with the Square Micro Fluorometer cells listed above, Black Anodized, Ext Width: 12.5mm, Ext Length: 12.5mm, Ext Height: 30mm, Int Width: 5.55mm, Int Length: 5.55mm, Aperature Width: 2.5mm, Aperature Height: 11mm