Learn More
AVANTOR MASTERFLEX (VWR LLC) Adapter, nylon, male luer to 1/8-27 thread, 25/pack
Catalog No. NC0595364
Encompass
Adapter, nylon, male luer to 1/8-27 thread, 25/pack. These adapters transition between luer fittings and UNF or NPT threads. All threaded connections are male thread.
Catalog No. NC0595364 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4550586
Adapter, nylon, male luer to 1/8-27 thread, 25/pack. These adapters transition between luer fittings and UNF or NPT threads. All threaded connections are male thread.