Learn More
Chemglass Life Sciences Adapter, Connecting, 14/20 Joints, 30mm Between Joints, Straight
SDP
Supplier:  Chemglass Life Sciences CG100709
Encompass_Preferred
Adapter, Connecting, 14/20 Joints, 30mm Between Joints, Straight. Used to expand the length of apparatus. Adapter can also be used as an air condenser. With standard taper top outer and lower inner joint.
Catalog No. CG100709