Learn More
Chemglass Life Sciences Adapter, Connecting, 24/40 Joints, 70mm Between Joints, Straight
SDP
Catalog No. CG100702
Encompass_Preferred
Adapter, Connecting, 24/40 Joints, 70mm Between Joints, Straight. Used to expand the length of apparatus. Adapter can also be used as an air condenser. With standard taper top outer and lower inner joint.
Catalog No. CG100702 Supplier Chemglass Life Sciences Supplier No. CG100702
Adapter, Connecting, 24/40 Joints, 70mm Between Joints, Straight. Used to expand the length of apparatus. Adapter can also be used as an air condenser. With standard taper top outer and lower inner joint.