Learn More
Aviva Systems Biology Corp ADARB1 Antibody - N-terminal region (ARP40342_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40342_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ADARB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADARB1. Alternate names: RED1, ADAR2, DRABA2, DRADA2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57400