Learn More
Aviva Systems Biology Corp ADARB1 Antibody - N-terminal region (ARP40343_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40343_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADARB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ADARB1. Alternate names: ADAR2, ADAR2a, ADAR2a-L1, ADAR2a-L2, ADAR2a-L3, ADAR2b, ADAR2d, ADAR2g, DRABA2, DRADA2, RED1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-57401