Learn More
Aviva Systems Biology Corp ADARB2 Antibody - middle region (ARP40918_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40918_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADARB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ADARB2. Alternate names: ADAR3, FLJ25034, FLJ36975, RED2, hRED2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57663