Learn More
TARGETMOL CHEMICALS INC Adatanserin hydrochloride 1MG
SDP
Catalog No. 502878927
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Adatanserin hydrochloride (WY50324 hydrochloride) is a novel 5-HT(1A)/5-HT(2) receptor ligand with potential neuroprotective effects and inhibition of ischemic efflux of endogenous amino acids, which can be used in the study of depression and anxiety disorders. Purity 99.64%
Catalog No. 50-287-8927 Supplier TARGETMOL CHEMICALS INC Supplier No. T296451MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Adatanserin hydrochloride (WY50324 hydrochloride) is a novel 5-HT(1A)/5-HT(2) receptor ligand with potential neuroprotective effects and inhibition of ischemic efflux of endogenous amino acids, which can be used in the study of depression and anxiety disorders. Purity 99.64%