Learn More
Medchemexpress LLC HY-10993 10mg Medchemexpress, Adavosertib CAS:955365-80-7 Purity:>98%
SDP
Catalog No. 502028802
Encompass_Preferred
Medchemexpress, HY-10993 10mg Adavosertib CAS:955365-80-7 Adavosertib (AZD-1775; MK-1775) is a potent Wee1 inhibitor with an IC50 of 5.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8802 Supplier Medchemexpress LLC Supplier No. HY1099310MG
Medchemexpress, HY-10993 10mg Adavosertib CAS:955365-80-7 Adavosertib (AZD-1775; MK-1775) is a potent Wee1 inhibitor with an IC50 of 5.2 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.