Learn More
Zymo Research Corporation ADB (Agarose Dissolving Buffer), 50 ml
SDP
Catalog No. 50444176
Encompass_Preferred
This buffer dissolves agarose gel slices for the recovery of DNA and facilitates efficient DNA binding and subsequent recovery.
Catalog No. 50-444-176 Supplier Zymo Research Corporation Supplier No. D4001150
This buffer dissolves agarose gel slices for the recovery of DNA and facilitates efficient DNA binding and subsequent recovery.