Learn More
MSC ADB Hoist Rings Center Pull Hoist Ring, 2500 lb Capacity, 1/2-13 Thread
Catalog No. 5000528579
Encompass
This Center Pull Hoist Ring is designed for secure lifting applications with a robust screw-on attachment. It features a substantial load capacity of 2500 lbs and a 1/2-13 thread size for compatibility with common fastening systems. Constructed from black oxide alloy steel, it offers enhanced durability and corrosion resistance for a longer service life.
  • Reliable screw-on attachment
  • 2500 lb load capacity
  • 1/2-13 thread size for broad compatibility
  • Black oxide alloy steel construction for enhanced durability and corrosion resistance
  • 4.12 in ring opening
  • 0.75 in bail diameter
  • 0.75 in thread length
  • Manufactured in the United States
Catalog No. 50-005-28579 Supplier MSC Supplier No. 48716039
This Center Pull Hoist Ring is designed for secure lifting applications with a robust screw-on attachment. It features a substantial load capacity of 2500 lbs and a 1/2-13 thread size for compatibility with common fastening systems. Constructed from black oxide alloy steel, it offers enhanced durability and corrosion resistance for a longer service life.
  • Reliable screw-on attachment
  • 2500 lb load capacity
  • 1/2-13 thread size for broad compatibility
  • Black oxide alloy steel construction for enhanced durability and corrosion resistance
  • 4.12 in ring opening
  • 0.75 in bail diameter
  • 0.75 in thread length
  • Manufactured in the United States