Learn More
MSC ADB Hoist Rings Center Pull Hoist Ring, 450 kg, M10 x 1.5, Black Oxide Alloy Steel
Catalog No. 5000530042
Encompass
This hoist ring features a center pull design for screw-on applications. It has a load capacity of 450 kg and uses an M10 x 1.5 thread. Constructed from black oxide alloy steel, it is designed for secure lifting and rigging operations.
  • Center pull type
  • 450 kg load capacity
  • M10 x 1.5 thread size
  • Black oxide alloy steel material
  • 82.00 mm ring height
  • 16.00 mm thread length
  • 31.80 mm mounting surface diameter
  • 16.50 mm radius
  • Manufactured in the United States
Catalog No. 50-005-30042 Supplier MSC Supplier No. 00950246
This hoist ring features a center pull design for screw-on applications. It has a load capacity of 450 kg and uses an M10 x 1.5 thread. Constructed from black oxide alloy steel, it is designed for secure lifting and rigging operations.
  • Center pull type
  • 450 kg load capacity
  • M10 x 1.5 thread size
  • Black oxide alloy steel material
  • 82.00 mm ring height
  • 16.00 mm thread length
  • 31.80 mm mounting surface diameter
  • 16.50 mm radius
  • Manufactured in the United States