Learn More
MSC ADB Hoist Rings Center Pull Hoist Ring, 5000 lb Capacity, 3/4-10 Thread, Black Oxide Alloy Steel
Catalog No. 5000533299
Encompass
This center pull hoist ring is designed for lifting applications requiring sling angle adjustments. It features a 5000 lb load capacity and a 3/4-10 thread size, constructed from black oxide alloy steel for durability.
  • Allows for sling angle adjustments
  • 5000 lb load capacity
  • 3/4-10 thread size
  • Black oxide alloy steel construction
Catalog No. 50-005-33299 Supplier MSC Supplier No. 06921860
This center pull hoist ring is designed for lifting applications requiring sling angle adjustments. It features a 5000 lb load capacity and a 3/4-10 thread size, constructed from black oxide alloy steel for durability.
  • Allows for sling angle adjustments
  • 5000 lb load capacity
  • 3/4-10 thread size
  • Black oxide alloy steel construction