Learn More
MSC ADB Hoist Rings Center Pull Hoist Ring, 800 lb Capacity, 5/16-18 Thread
Catalog No. 5000529386
Encompass
This center pull hoist ring is designed for secure lifting applications. It features an 800 lb capacity and a 5/16-18 thread for reliable attachment. Constructed from black oxide alloy steel, it offers durability and strength.
  • 800 lb load capacity
  • 5/16-18 thread size
  • Black oxide alloy steel construction
  • 1.51 in ring opening
  • 2.29 in ring width
  • 1.06 in thread length
  • 1.25 in mounting surface diameter
  • 7 ft/lb torque rating
  • Made in the United States
Catalog No. 50-005-29386 Supplier MSC Supplier No. 06921720
This center pull hoist ring is designed for secure lifting applications. It features an 800 lb capacity and a 5/16-18 thread for reliable attachment. Constructed from black oxide alloy steel, it offers durability and strength.
  • 800 lb load capacity
  • 5/16-18 thread size
  • Black oxide alloy steel construction
  • 1.51 in ring opening
  • 2.29 in ring width
  • 1.06 in thread length
  • 1.25 in mounting surface diameter
  • 7 ft/lb torque rating
  • Made in the United States