Learn More
MSC ADB Hoist Rings Center Pull Hoist Ring, 800 lb Capacity, 5/16-18 Thread
Catalog No. 5000529449
Encompass
This center pull hoist ring is designed for lifting applications, offering an 800 lb capacity. It is constructed from durable alloy steel with a black oxide finish and features a 5/16-18 thread for secure attachment.
  • Screw-on design
  • 800 lb load capacity
  • 5/16-18 thread size
  • Alloy steel construction
  • Black oxide finish
  • 1.51 in ring opening
  • 2.29 in ring width
  • 0.56 in thread length
  • 1.25 in mounting surface diameter
  • 7.000 ft-lb torque
Catalog No. 50-005-29449 Supplier MSC Supplier No. 06921704
This center pull hoist ring is designed for lifting applications, offering an 800 lb capacity. It is constructed from durable alloy steel with a black oxide finish and features a 5/16-18 thread for secure attachment.
  • Screw-on design
  • 800 lb load capacity
  • 5/16-18 thread size
  • Alloy steel construction
  • Black oxide finish
  • 1.51 in ring opening
  • 2.29 in ring width
  • 0.56 in thread length
  • 1.25 in mounting surface diameter
  • 7.000 ft-lb torque