Learn More
MSC ADB Hoist Rings Center Pull Hoist Ring, 8000 lb Capacity, 7/8-9 Thread
Catalog No. 5000529533
Encompass
This center pull hoist ring features a screw-on design and a robust load capacity of 8000 lb. It is constructed from black oxide alloy steel and has a 7/8-9 thread size, ideal for secure lifting applications. Key specifications include a ring opening of 3.32 in and a thread length of 1.20 in, with a torque rating of 160 ft/lb.
  • Screw-on design for easy installation
  • 8000 lb load capacity for heavy-duty lifting
  • Black oxide alloy steel construction for durability
  • 7/8-9 thread size ensures secure attachment
  • 3.32 in ring opening provides ample space
  • 1.20 in thread length for reliable mounting
  • 160 ft/lb torque rating for secure fastening
Catalog No. 50-005-29533 Supplier MSC Supplier No. 06921928
This center pull hoist ring features a screw-on design and a robust load capacity of 8000 lb. It is constructed from black oxide alloy steel and has a 7/8-9 thread size, ideal for secure lifting applications. Key specifications include a ring opening of 3.32 in and a thread length of 1.20 in, with a torque rating of 160 ft/lb.
  • Screw-on design for easy installation
  • 8000 lb load capacity for heavy-duty lifting
  • Black oxide alloy steel construction for durability
  • 7/8-9 thread size ensures secure attachment
  • 3.32 in ring opening provides ample space
  • 1.20 in thread length for reliable mounting
  • 160 ft/lb torque rating for secure fastening