Learn More
MSC ADB Hoist Rings Center Pull Hoist Ring, Screw-On, 10000 lb Capacity, 1-8 Thread
Catalog No. 5000529070
Encompass
This Center Pull Hoist Ring is designed for screw-on applications, offering a robust 10000 lb load capacity. It features a 1-8 thread and is constructed from durable black oxide alloy steel.
  • Center pull type for versatile lifting
  • Screw-on mounting for secure attachment
  • 10000 lb load capacity
  • 1-8 thread for strong connection
  • Black oxide alloy steel construction
  • 3.20 in ring opening
  • 5.10 in ring width
  • 2.20 in thread length
  • 2.81 in mounting surface diameter
  • 230 ft/lb torque
Catalog No. 50-005-29070 Supplier MSC Supplier No. 06921951
This Center Pull Hoist Ring is designed for screw-on applications, offering a robust 10000 lb load capacity. It features a 1-8 thread and is constructed from durable black oxide alloy steel.
  • Center pull type for versatile lifting
  • Screw-on mounting for secure attachment
  • 10000 lb load capacity
  • 1-8 thread for strong connection
  • Black oxide alloy steel construction
  • 3.20 in ring opening
  • 5.10 in ring width
  • 2.20 in thread length
  • 2.81 in mounting surface diameter
  • 230 ft/lb torque