Learn More
MSC ADB Hoist Rings Center Pull Hoist Ring, Screw-On, 2200 kg, M20 x 2.5
Catalog No. 5000513582
Encompass
This center pull hoist ring is designed for secure lifting applications. It features a screw-on mechanism, a robust 2200 kg load capacity, and an M20 x 2.5 thread, ensuring strong and reliable attachment. The hoist ring is constructed from black oxide alloy steel for enhanced durability.
  • Screw-on type for easy installation
  • 2200 kg load capacity for heavy-duty lifting
  • M20 x 2.5 thread size for compatible mounting
  • Constructed from black oxide alloy steel for durability
  • Ring height of 134.90 mm
  • Thread length of 25.00 mm
Catalog No. 50-005-13582 Supplier MSC Supplier No. 00950303
This center pull hoist ring is designed for secure lifting applications. It features a screw-on mechanism, a robust 2200 kg load capacity, and an M20 x 2.5 thread, ensuring strong and reliable attachment. The hoist ring is constructed from black oxide alloy steel for enhanced durability.
  • Screw-on type for easy installation
  • 2200 kg load capacity for heavy-duty lifting
  • M20 x 2.5 thread size for compatible mounting
  • Constructed from black oxide alloy steel for durability
  • Ring height of 134.90 mm
  • Thread length of 25.00 mm