Learn More
MSC ADB Hoist Rings Hoist Ring, 450 kg, M10 x 1.5, Black Oxide Alloy Steel
Catalog No. 5000534188
Encompass
This center pull hoist ring is designed for secure lifting applications, featuring a 450 kg load capacity and an M10x1.5 thread. Constructed from black oxide alloy steel, it provides both strength and corrosion resistance. Its design allows for a 180° pivot and 360° swivel, enabling flexible load positioning.
  • 450 kg load capacity for heavy-duty applications.
  • M10 x 1.5 thread size for secure installation.
  • Alloy steel construction with black oxide finish provides durability and corrosion resistance.
  • Features 180° pivot and 360° swivel for flexible load handling.
  • Engineered with a minimum design factor of 5:1.
  • Magnetic particle inspected per ASTM E1444 and 200% proof load tested.
  • Meets ANSI/ASME B30.26 and Military Specification No. MIL-STD 209.
Catalog No. 50-005-34188 Supplier MSC Supplier No. 00950253
This center pull hoist ring is designed for secure lifting applications, featuring a 450 kg load capacity and an M10x1.5 thread. Constructed from black oxide alloy steel, it provides both strength and corrosion resistance. Its design allows for a 180° pivot and 360° swivel, enabling flexible load positioning.
  • 450 kg load capacity for heavy-duty applications.
  • M10 x 1.5 thread size for secure installation.
  • Alloy steel construction with black oxide finish provides durability and corrosion resistance.
  • Features 180° pivot and 360° swivel for flexible load handling.
  • Engineered with a minimum design factor of 5:1.
  • Magnetic particle inspected per ASTM E1444 and 200% proof load tested.
  • Meets ANSI/ASME B30.26 and Military Specification No. MIL-STD 209.