Learn More
MSC ADB Hoist Rings Hoist Ring, Center Pull, Screw-On, 5000 lb Capacity, Alloy Steel
Catalog No. 5000513226
Encompass
This hoist ring is designed for secure lifting applications, featuring a center pull mechanism and a screw-on attachment. It provides a robust load capacity of 5000 lb and is manufactured from durable alloy steel, finished with black oxide for enhanced resistance.
  • Center pull design
  • Screw-on attachment type
  • 5000 lb load capacity
  • Durable alloy steel construction
  • Black oxide finish
  • 3/4-10 thread size
  • 4.78 inch ring height
  • 1.03 inch thread length
  • Requires 100 FT/LB torque for proper installation
Catalog No. 50-005-13226 Supplier MSC Supplier No. 06921332
This hoist ring is designed for secure lifting applications, featuring a center pull mechanism and a screw-on attachment. It provides a robust load capacity of 5000 lb and is manufactured from durable alloy steel, finished with black oxide for enhanced resistance.
  • Center pull design
  • Screw-on attachment type
  • 5000 lb load capacity
  • Durable alloy steel construction
  • Black oxide finish
  • 3/4-10 thread size
  • 4.78 inch ring height
  • 1.03 inch thread length
  • Requires 100 FT/LB torque for proper installation