Learn More
MSC ADB Hoist Rings Hoist Ring, Flip-Flop, Screw-On, 2500 lb, 180°
Catalog No. 5000513437
Encompass
This hoist ring is designed for secure lifting applications, featuring a flip-flop design for 180° movement and screw-on mounting. It has a robust 2500 lb capacity and is constructed from forged chromium-molybdenum steel with a black oxide finish, ensuring durability and reliability for industrial use.
  • Flip-flop design allows for 180° movement during lifting operations.
  • Screw-on mount provides secure and reliable attachment.
  • Work load limit of 2500 lb for heavy-duty applications.
  • Constructed from forged chromium-molybdenum steel for exceptional strength and longevity.
  • Black oxide finish provides enhanced durability and corrosion resistance.
  • Meets ASTM 1444 standards for quality and safety.
  • Features a 3/8 thread size for versatile mounting.
Catalog No. 50-005-13437 Supplier MSC Supplier No. 00099473
This hoist ring is designed for secure lifting applications, featuring a flip-flop design for 180° movement and screw-on mounting. It has a robust 2500 lb capacity and is constructed from forged chromium-molybdenum steel with a black oxide finish, ensuring durability and reliability for industrial use.
  • Flip-flop design allows for 180° movement during lifting operations.
  • Screw-on mount provides secure and reliable attachment.
  • Work load limit of 2500 lb for heavy-duty applications.
  • Constructed from forged chromium-molybdenum steel for exceptional strength and longevity.
  • Black oxide finish provides enhanced durability and corrosion resistance.
  • Meets ASTM 1444 standards for quality and safety.
  • Features a 3/8 thread size for versatile mounting.