Learn More
MSC ADB Hoist Rings Safety Engineered Hoist Ring, 8000 lb, 7/8-9 Thread, Black Oxide 4140 Alloy Steel
Catalog No. 5000529669
Encompass
ADB Hoist Rings Safety Engineered Hoist Ring is a screw-on type hoist ring designed for heavy-duty lifting applications. It features an 8000 lb load capacity, a 7/8-9 thread, and is constructed from black oxide 4140 alloy steel for corrosion resistance and durability.
  • Screw-on design for secure attachment
  • 8000 lb load capacity
  • 7/8-9 thread for robust connection
  • High strength 4140 alloy steel construction
  • Black oxide finish for corrosion resistance
  • Magnetic particle inspected per ASTM E1444
  • Retains rated load from 0 to 90 degrees from the bolt axis
  • Swivels 360 degrees and pivots 180 degrees under load
  • 200% proof load tested
  • Individually serial numbered
Catalog No. 50-005-29669 Supplier MSC Supplier No. 00545947
ADB Hoist Rings Safety Engineered Hoist Ring is a screw-on type hoist ring designed for heavy-duty lifting applications. It features an 8000 lb load capacity, a 7/8-9 thread, and is constructed from black oxide 4140 alloy steel for corrosion resistance and durability.
  • Screw-on design for secure attachment
  • 8000 lb load capacity
  • 7/8-9 thread for robust connection
  • High strength 4140 alloy steel construction
  • Black oxide finish for corrosion resistance
  • Magnetic particle inspected per ASTM E1444
  • Retains rated load from 0 to 90 degrees from the bolt axis
  • Swivels 360 degrees and pivots 180 degrees under load
  • 200% proof load tested
  • Individually serial numbered