Learn More
MSC ADB Hoist Rings Side Pull Hoist Ring, 3000 kg Capacity, M20 x 2.5 Thread
Catalog No. 5000527822
Encompass
This side pull hoist ring is designed for lifting applications requiring a high load capacity. It features a robust design suitable for heavy-duty tasks, ensuring secure attachment and safe handling of materials.
  • 3000 kg capacity
  • M20 x 2.5 thread size
  • Clevis height of 4-3/16 in
  • Clevis width of 3-3/4 in
  • Width at screw of 5-1/4 in
  • Thread length of 39.00 mm
  • Ring thickness of 19.00 mm
  • Radius of 22.00 mm
  • Manufactured in the United States
Catalog No. 50-005-27822 Supplier MSC Supplier No. 48724215
This side pull hoist ring is designed for lifting applications requiring a high load capacity. It features a robust design suitable for heavy-duty tasks, ensuring secure attachment and safe handling of materials.
  • 3000 kg capacity
  • M20 x 2.5 thread size
  • Clevis height of 4-3/16 in
  • Clevis width of 3-3/4 in
  • Width at screw of 5-1/4 in
  • Thread length of 39.00 mm
  • Ring thickness of 19.00 mm
  • Radius of 22.00 mm
  • Manufactured in the United States