Learn More
Cayman Chemical ADBICA NpentnoIc ACD metab 5mg
SDP
Catalog No. 502971898
Encompass_Preferred
A potential phase 1 metabolite of ADBICA, the synthetic CB recently identified in herbal blends; intended for forensic and research applications
Catalog No. 50-297-1898 Supplier Cayman Chemical Supplier No. 150615 MG
A potential phase 1 metabolite of ADBICA, the synthetic CB recently identified in herbal blends; intended for forensic and research applications