Learn More
Cayman Chemical ADBPINCA pentnoIc ACD met 10mg
SDP
Catalog No. 502969499
Encompass_Preferred
An expected compound produced by phase I metabolism of ADB-PINACA in vivo, based on the metabolism of comparable alkylindole CBs
Catalog No. 50-296-9499 Supplier Cayman Chemical Supplier No. 1505510 MG
An expected compound produced by phase I metabolism of ADB-PINACA in vivo, based on the metabolism of comparable alkylindole CBs