Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Adbrettphos Pd G3

Catalog No. 7761062G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
77-610-62G 5 g
77-610-65G 500 mg
77-610-6500MG 100 mg
77-610-6100MG 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 77-610-62G Supplier Sigma Aldrich Supplier No. 7761062G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Adbrettphos Pd G3

Specifications

CAS 72320-38-8
Density 1.095 g/mL at 25°C
Molecular Formula C3H7N3O
Linear Formula C3H7N3O
Refractive Index n20/D 1.461
MDL Number MFCD14652506
Quantity 5 g
Molecular Weight (g/mol) 101.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.