Learn More
AVANTOR MASTERFLEX (VWR LLC) ADCF Female Luer to 1/8"" J Barb Adapter, PP, 25/pk
Catalog No. NC0812921
Encompass
ADCF Female Luer to 1/8" J Barb Adapter, Polypropylene, 25/pk, Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving). Sold in packs of 25.
Catalog No. NC0812921 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3080002
ADCF Female Luer to 1/8" J Barb Adapter, Polypropylene, 25/pk, Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving). Sold in packs of 25.