Learn More
Aviva Systems Biology Corp ADCK2 Antibody - C-terminal region (ARP63132_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63132_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADCK2. It was validated on Western Blot. . Alternate names: AARF, MGC20727ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-891