Learn More
Sino Biological Recombinant Human DGKQ Protein (Full Length, His Tag), Active
Catalog No. 502575337
Encompass_Preferred
Recombinant full length human DGKQ was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-5337 Supplier Sino Biological Supplier No. D2910H50
Recombinant full length human DGKQ was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.