Learn More
Aviva Systems Biology Corp ADCY2 Antibody - middle region (ARP49526_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADCY2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ADCY2. Alternate names: AC2, FLJ16822, FLJ45092, HBAC2, KIAA1060, MGC133314ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-285-434