Learn More
Aviva Systems Biology Corp Adcyap1r1 Antibody - C-terminal region (ARP59945_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59945_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Adcyap1r1. It was validated on Western Blot. . Alternate names: PACAP-R1A, PACAPR1|PACAPR1A, PAC1-R, PACAPR1, PACAPR1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-887