Learn More
Chemglass Life Sciences 250mL Addition Funnel, Cooling Jacket, 24/40 Joint Size, 2mm Stpk
SDP
Catalog No. CG170305
Encompass_Preferred
250mL Addition Funnel, Cooling Jacket, 24/40 Joint Size, 2mm Stopcock. Jacket permits the cooling of the contents of the funnel by means of dry ice. Drain outlet is 8mm O.D. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm.
Catalog No. CG170305 Supplier Chemglass Life Sciences Supplier No. CG170305
250mL Addition Funnel, Cooling Jacket, 24/40 Joint Size, 2mm Stopcock. Jacket permits the cooling of the contents of the funnel by means of dry ice. Drain outlet is 8mm O.D. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm.