Learn More
VAZYME INTERNATIONAL LLC Vazyme Add&Read TR-FRET cAMP Detection Kit, 1000 tests
Supplier:  VAZYME INTERNATIONAL LLC DD280101
Encompass
The Add&Read TR-FRET cAMP Detection Kit is a no-wash ELISA-based solution designed for cAMP detection, offering significant time and labor savings compared to traditional ELISA methods. This kit is configured for 1000 tests.
  • Based on TR-FRET methodology.
  • Utilizes a “no wash elisa” approach.
  • Reduces time and labor by over 80% compared to traditional ELISA due to its two-step process.
Catalog No. NC3979682