Learn More
Aviva Systems Biology Corp ADD3 Antibody - middle region (ARP59947_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59947_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADD3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ADD3. Alternate names: ADDLApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-889