Learn More
Medchemexpress LLC HY-U00448 5mg Medchemexpress, ADDA 5 hydrochloride CAS:473268-46-1 Purity:>98%
SDP
Catalog No. 501934456
Encompass_Preferred
Medchemexpress, HY-U00448 5mg ADDA 5 hydrochloride CAS:473268-46-1 ADDA 5 hydrochloride is a partial non-competitive inhibitor of cytochrome c oxidase (CcO) , with IC 50 s of 18.93 μM and 31.82 μM for purified CcO from human glioma and bovine heart, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4456 Supplier Medchemexpress LLC Supplier No. HYU004485MG
Medchemexpress, HY-U00448 5mg ADDA 5 hydrochloride CAS:473268-46-1 ADDA 5 hydrochloride is a partial non-competitive inhibitor of cytochrome c oxidase (CcO) , with IC 50 s of 18.93 μM and 31.82 μM for purified CcO from human glioma and bovine heart, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.