Learn More
Chemglass Life Sciences 125mL Addition Funnel, 24/40 Joint Size, 2mm PTFE Stpk, 325mm OAH
SDP
Catalog No. CG170202
Encompass_Preferred
125mL Addition Funnel, 24/40 Joint Size, 2mm PTFE Stopcock, 325mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.
Catalog No. CG170202 Supplier Chemglass Life Sciences Supplier No. CG170202
125mL Addition Funnel, 24/40 Joint Size, 2mm PTFE Stopcock, 325mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.