Learn More
Ace Glass, Inc. Addition funnel, 19L, #25 Ace-Thred bottom, body only
SDP
Catalog No. 50974742
Encompass
19L addition/separatory funnel body for use where large volumes are needed. The cylindrical shape necks down to a #25 Ace-Thred.
Catalog No. 50-974-742 Supplier Ace Glass, Inc. Supplier No. 727210
19L addition/separatory funnel body for use where large volumes are needed. The cylindrical shape necks down to a #25 Ace-Thred.