Learn More
Chemglass Life Sciences 250mL Addition Funnel, 24/40 Joint Size, 2mm PTFE Stpk, 375mm OAH
SDP
Catalog No. CG170203
Encompass_Preferred
250mL Addition Funnel, 24/40 Joint Size, 2mm PTFE Stopcock, 375mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.
Catalog No. CG170203 Supplier Chemglass Life Sciences Supplier No. CG170203
250mL Addition Funnel, 24/40 Joint Size, 2mm PTFE Stopcock, 375mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and PTFE stopcock. For replacement stoppers see CG-3000.