Learn More
Ace Glass, Inc. Addition funnel, 35L, complete with #25 PTFE coupling and8mm bottom glass stopcock adapter with PTFE plug
SDP
Catalog No. 50979236
Encompass
35L addition/separatory funnel for use where large volumes are needed. The cylindrical shape necks down to a #25 Ace-Thred, and connects to an 8mm bore PTFE-clad glass stopcock via a #25 PTFE coupling using FETFE O-rings. The funnel system is completely grease-free. Funnel system includes funnel body, coupling with two O-rings, and a stopcock.
Catalog No. 50-979-236 Supplier Ace Glass, Inc. Supplier No. 727244
35L addition/separatory funnel for use where large volumes are needed. The cylindrical shape necks down to a #25 Ace-Thred, and connects to an 8mm bore PTFE-clad glass stopcock via a #25 PTFE coupling using FETFE O-rings. The funnel system is completely grease-free. Funnel system includes funnel body, coupling with two O-rings, and a stopcock.