Learn More
Chemglass Life Sciences 50mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stpk, 270mm OAH
SDP
Catalog No. CG171818
Encompass_Preferred
50mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stpk, 270mm OAH. Cylindrical style addition funnel similar to CG-1716, but with PTFE stopcocks. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171818 Supplier Chemglass Life Sciences Supplier No. CG171818
50mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Stpk, 270mm OAH. Cylindrical style addition funnel similar to CG-1716, but with PTFE stopcocks. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.