Learn More
Ace Glass, Inc. Addition funnel, 9.5L, #25 Ace-Thred bottom, body only
SDP
Catalog No. 50979235
Encompass
9.5L addition/separatory funnel body for use where large volumes are needed. The cylindrical shape necks down to a #25 Ace-Thred.
Catalog No. 50-979-235 Supplier Ace Glass, Inc. Supplier No. 727204
Add to Cart
Add to Cart
9.5L addition/separatory funnel body for use where large volumes are needed. The cylindrical shape necks down to a #25 Ace-Thred.