Learn More
Chemglass Life Sciences 125mL Addition Funnel, 24/40 Joint Size, 0-2mm PTFE Valve, 350mm OAH
SDP
Catalog No. CG170402
Encompass_Preferred
125mL Addition Funnel, 24/40 Joint Size, 0-2mm PTFE Valve, 350mm OAH. Cylindrical style addition funnel with a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. For replacement stoppers see CG-3000.
Catalog No. CG170402 Supplier Chemglass Life Sciences Supplier No. CG170402
125mL Addition Funnel, 24/40 Joint Size, 0-2mm PTFE Valve, 350mm OAH. Cylindrical style addition funnel with a threaded, grease-free extended tip valve. Design of all PTFE plug permits semi-needle valve action. With a standard taper top outer joint and lower inner joint, both being the same size, and pressure equalizing arm. For replacement stoppers see CG-3000.