Learn More
Bellco Glass, Inc. Additional/Vent Pipe Assembly For 10mm Port
SDP
Catalog No. 501946439
Encompass_Preferred
316 L stainless steel addition port with hose barb for attachment of silicone tubing. This can be used for the introduction of small volumes aseptically into the bioreactor vessel.. MADE IN THE USA
Catalog No. 50-194-6439 Supplier Bellco Glass, Inc. Supplier No. 790900028
316 L stainless steel addition port with hose barb for attachment of silicone tubing. This can be used for the introduction of small volumes aseptically into the bioreactor vessel.. MADE IN THE USA