Learn More
XONA MICROFLUIDICS INC XC Pre-Coat™ solution
SDP
Catalog No. NC3316442
Encompass
Additional XC Pre-Coat™ solutionEach Box of (5) XC-Series uses (1) 2 ml Tube of XC Pre-coat™Each Box of (5) XC-T Series uses (2) 2 ml Tubes of XC Pre-coat™
Catalog No. NC3316442 Supplier XONA MICROFLUIDICS INC Supplier No. XC PRECOAT
Add to Cart
Add to Cart
Additional XC Pre-Coat™ solutionEach Box of (5) XC-Series uses (1) 2 ml Tube of XC Pre-coat™Each Box of (5) XC-T Series uses (2) 2 ml Tubes of XC Pre-coat™