Learn More
Chemglass Life Sciences 60mL Addition Funnel, 14/20 Joint Size, 2mm Glass Stpk, 220mm OAH
SDP
Catalog No. CG170016
Encompass_Preferred
60mL Addition Funnel, 14/20 Joint Size, 2mm Glass Stopcock, 220mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG170016 Supplier Chemglass Life Sciences Supplier No. CG170016
60mL Addition Funnel, 14/20 Joint Size, 2mm Glass Stopcock, 220mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.