Learn More
Takara Bio ADE/ HIS/ LEU/ TRP SUPPLMNT
SDP
Catalog No. NC9530091
Encompass
Protein Interaction, -Ade/ -His/ -Leu/ -Trp DO Supplement, 10g, For two-hybrid library screening using the Pretransformed Matchmaker cDNA Libraries
Catalog No. NC9530091 Supplier Takara Bio Supplier No. 630428
Protein Interaction, -Ade/ -His/ -Leu/ -Trp DO Supplement, 10g, For two-hybrid library screening using the Pretransformed Matchmaker cDNA Libraries